MAT047025495 Matrix Scientific

4-Ethylphenylhydrazine hydrochloride

  • MDL number MFCD01861267
  • Cas Number : 53661-18-0
  • Molecular Weight : 172.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128-130
Country of Origin USA
Product Link http://www.matrixscientific.com/007270.html

Safety Information

hcodes Irritant/Irritant-Harmful
MSDS http://www.matrixscientific.com/007270.html
Availability details for
Quantity
  • Availability
  • Add to Cart