MAT047025507 Matrix Scientific

2-(4-Fluorophenyl)malondialdehyde

  • MDL number MFCD02261897
  • Cas Number : 493036-47-8
  • Molecular Weight : 166.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-167
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/007281.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007281.html
Availability details for
Quantity
  • Availability
  • Add to Cart