MAT047025508 Matrix Scientific

2-(4-Trifluoromethylphenyl)malondialdehyde

  • MDL number MFCD03424497
  • Cas Number : 493036-50-3
  • Molecular Weight : 216.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-134
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/007282.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007282.html
Availability details for
Quantity
  • Availability
  • Add to Cart