MAT047025510 Matrix Scientific

2-(3-Chlorophenyl)malonaldehyde

  • MDL number MFCD02261887
  • Cas Number : 74963-16-9
  • Molecular Weight : 182.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107-109
Country of Origin USA
Product Link http://www.matrixscientific.com/007285.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007285.html
Availability details for
Quantity
  • Availability
  • Add to Cart