MAT047025515 Matrix Scientific

1-Phenyl-2-(pyrazin-2-yl)ethanol

  • MDL number MFCD00235147
  • Cas Number : 36914-69-9
  • Molecular Weight : 200.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88-90
Country of Origin USA
Product Link http://www.matrixscientific.com/007289.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007289.html
Availability details for
Quantity
  • Availability
  • Add to Cart