MAT047025524 Matrix Scientific

1-(4-Methoxyphenyl)-2-(quinolin-2-yl)ethanol

  • MDL number MFCD03452779
  • Cas Number : 94004-77-0
  • Molecular Weight : 279.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-140
Country of Origin USA
Product Link http://www.matrixscientific.com/007302.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007302.html
Availability details for
Quantity
  • Availability
  • Add to Cart