MAT047025525 Matrix Scientific

2-Chloro-5-(4-methoxyphenyl)pyrimidine

  • MDL number MFCD03426059
  • Cas Number : 27794-03-2
  • Molecular Weight : 220.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129-131
Country of Origin USA
Product Link http://www.matrixscientific.com/007303.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007303.html
Availability details for
Quantity
  • Availability
  • Add to Cart