MAT047025535 Matrix Scientific

1-[(2-Methoxyethyl)amino]-1-(4-methylsulfonylphenyl)ethane

  • MDL number MFCD00203232
  • Cas Number : 849924-92-1
  • Molecular Weight : 257.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007312.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007312.html
Availability details for
Quantity
  • Availability
  • Add to Cart