MAT047025540 Matrix Scientific

1,2-Dimethyl-1H-indole-3-carboxaldehyde

  • MDL number MFCD00745294
  • Cas Number : 38292-40-9
  • Molecular Weight : 173.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127-129
Country of Origin USA
Boiling point 200-205�Torr
Product Link http://www.matrixscientific.com/007317.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007317.html
Availability details for
Quantity
  • Availability
  • Add to Cart