MAT047025552 Matrix Scientific

3-[3-(Pyrazin-2-yl)-1,2,4-oxadiazol-5-yl]-propanoic acid

  • MDL number MFCD03426086
  • Cas Number : 849925-05-9
  • Molecular Weight : 220.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >155�c)
Country of Origin USA
Product Link http://www.matrixscientific.com/007328.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007328.html
Availability details for
Quantity
  • Availability
  • Add to Cart