MAT047025554 Matrix Scientific

4-[3-(2-Thienyl)-1,2,4-oxadiazol-5-yl]-butanoic acid

  • MDL number MFCD03426087
  • Cas Number : 849925-06-0
  • Molecular Weight : 238.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87-90
Country of Origin USA
Product Link http://www.matrixscientific.com/007330.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007330.html
Availability details for
Quantity
  • Availability
  • Add to Cart