MAT047025558 Matrix Scientific

1-(5-Bromopyrimidin-2-yl)-[1,4]-diazepane

  • MDL number MFCD03426069
  • Cas Number : 849021-44-9
  • Molecular Weight : 257.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203-207
Country of Origin USA
Product Link http://www.matrixscientific.com/007333.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007333.html
Availability details for
Quantity
  • Availability
  • Add to Cart