MAT047025561 Matrix Scientific

2-Chloro-5-(4-chlorophenyl)pyrimidine

  • MDL number MFCD03426061
  • Cas Number : 27956-40-7
  • Molecular Weight : 225.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209-212�b)
Country of Origin USA
Product Link http://www.matrixscientific.com/007336.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007336.html
Availability details for
Quantity
  • Availability
  • Add to Cart