MAT047025574 Matrix Scientific

4-[3-(Diphenylmethyl)-1,2,4-oxadiazol-5-yl]-piperidine

  • MDL number MFCD03426081
  • Cas Number : 849925-02-6
  • Molecular Weight : 319.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-67
Country of Origin USA
Product Link http://www.matrixscientific.com/007350.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007350.html
Availability details for
Quantity
  • Availability
  • Add to Cart