MAT047025575 Matrix Scientific

4-[5-(4-Fluorophenyl)-1,3,4-oxadiazol-2-yl]-piperidine

  • MDL number MFCD03424488
  • Cas Number : 493024-40-1
  • Molecular Weight : 247.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93-96
Country of Origin USA
Product Link http://www.matrixscientific.com/007351.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007351.html
Availability details for
Quantity
  • Availability
  • Add to Cart