MAT047025576 Matrix Scientific

4-[(2-Methoxyethyl)amino]-1-methylpiperidine

  • MDL number MFCD01826433
  • Cas Number : 416887-38-2
  • Molecular Weight : 172.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 62-64�03mm
Product Link http://www.matrixscientific.com/007352.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007352.html
Availability details for
Quantity
  • Availability
  • Add to Cart