MAT047025580 Matrix Scientific

5-[5-(Piperidin-4-yl)-1,2,4-oxadiazol-3-yl]-1,3-benzodioxole

  • MDL number MFCD03426084
  • Cas Number : 849925-04-8
  • Molecular Weight : 273.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93-96
Country of Origin USA
Product Link http://www.matrixscientific.com/007355.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007355.html
Availability details for
Quantity
  • Availability
  • Add to Cart