MAT047025582 Matrix Scientific

3-[5-(Piperidin-4-yl)-[1,2,4]-oxadiazol-3-yl]-pyridine

  • MDL number MFCD03426078
  • Cas Number : 849925-01-5
  • Molecular Weight : 230.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80-83
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/007357.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007357.html
Availability details for
Quantity
  • Availability
  • Add to Cart