MAT047025595 Matrix Scientific

2-(tert-Butoxymethyl)phenylboronic acid

  • MDL number MFCD03425930
  • Cas Number : 373384-12-4
  • Molecular Weight : 208.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin USA
Product Link http://www.matrixscientific.com/007374.html

Safety Information

hcodes Irritant/Store in Freezer
MSDS http://www.matrixscientific.com/007374.html
Availability details for
Quantity
  • Availability
  • Add to Cart