MAT047025603 Matrix Scientific

4-Iodo-2-methylanisole

  • MDL number MFCD03425972
  • Cas Number : 75581-11-2
  • Molecular Weight : 248.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75-76
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/007383.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007383.html
Availability details for
Quantity
  • Availability
  • Add to Cart