MAT047025608 Matrix Scientific

Pyrazine-2-carbothioic acid,S-phenyl ester

  • MDL number MFCD03425980
  • Cas Number : 121761-15-7
  • Molecular Weight : 216.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149-151
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/007388.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/007388.html
Availability details for
Quantity
  • Availability
  • Add to Cart