MAT047025670 Matrix Scientific

2-(Heptafluoro-n-propyl)benzimidazole

  • MDL number MFCD00521800
  • Cas Number : 559-37-5
  • Molecular Weight : 286.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225-226
Country of Origin USA
Product Link http://www.matrixscientific.com/007449.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007449.html
Availability details for
Quantity
  • Availability
  • Add to Cart