MAT047025671 Matrix Scientific

3-Aminobenzanilide

  • MDL number MFCD00017101
  • Cas Number : 14315-16-3
  • Molecular Weight : 212.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123-124
Purity 97
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/007450.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007450.html
Availability details for
Quantity
  • Availability
  • Add to Cart