MAT047025689 Matrix Scientific

2-Bromo-5-chlorophenylacetic acid

  • MDL number MFCD00153890
  • Cas Number : 81682-38-4
  • Molecular Weight : 249.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101-103
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/001647.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001647.html
Availability details for
Quantity
  • Availability
  • Add to Cart