MAT047025692 Matrix Scientific

4-Phenyl-1,2,3-thiadiazole

  • MDL number MFCD00087232
  • Cas Number : 25445-77-6
  • Molecular Weight : 162.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77-78
Purity 98
Country of Origin USA
Boiling point 100�5 Torr
Product Link http://www.matrixscientific.com/007470.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007470.html
Availability details for
Quantity
  • Availability
  • Add to Cart