MAT047025694 Matrix Scientific

4-Isopropylphenylacetic acid

  • MDL number MFCD00041035
  • Cas Number : 4476-28-2
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51-52
Purity 98
Country of Origin USA
Boiling point 170-174� Torr
Product Link http://www.matrixscientific.com/007472.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007472.html
Availability details for
Quantity
  • Availability
  • Add to Cart