MAT047025698 Matrix Scientific

4-Methoxy-2-methylphenylacetonitrile

  • MDL number MFCD02178388
  • Cas Number : 262298-02-2
  • Molecular Weight : 161.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/007476.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007476.html
Availability details for
Quantity
  • Availability
  • Add to Cart