MAT047025702 Matrix Scientific

2,4-Difluorobenzhydrazide

  • MDL number MFCD00173685
  • Cas Number : 118737-62-5
  • Molecular Weight : 172.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141-142
Country of Origin USA
Product Link http://www.matrixscientific.com/007015.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007015.html
Availability details for
Quantity
  • Availability
  • Add to Cart