MAT047025708 Matrix Scientific

5-(2,6-Difluorophenyl)isoxazole

  • MDL number MFCD02183540
  • Cas Number : 138716-41-3
  • Molecular Weight : 181.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007020.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007020.html
Availability details for
Quantity
  • Availability
  • Add to Cart