MAT047025740 Matrix Scientific

Ethyl 4-hydroxy-6-(trifluoromethyl)-3-quinolinecarboxylate

  • MDL number MFCD00173369
  • Cas Number : 26893-12-9
  • Molecular Weight : 285.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007022.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007022.html
Availability details for
Quantity
  • Availability
  • Add to Cart