MAT047025773 Matrix Scientific

N,N'-Di-tert-butyl-1,3-propanediamine

  • MDL number MFCD00196058
  • Cas Number : 22687-38-3
  • Molecular Weight : 186.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40
Purity 97
Country of Origin USA
Boiling point 97�8 Torr
Product Link http://www.matrixscientific.com/007553.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007553.html
Availability details for
Quantity
  • Availability
  • Add to Cart