MAT047025774 Matrix Scientific

2,6-Dichlorobenzylamine

  • MDL number MFCD00047928
  • Cas Number : 6575-27-5
  • Molecular Weight : 176.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40
Purity 97
Country of Origin USA
Boiling point 108�mm
Product Link http://www.matrixscientific.com/007554.html

Safety Information

hcodes Corrosive/Air Sensitive
MSDS http://www.matrixscientific.com/007554.html
Availability details for
Quantity
  • Availability
  • Add to Cart