MAT047025779 Matrix Scientific

5-(Diethylamino)pentyl alcohol

  • MDL number MFCD00046024
  • Cas Number : 2683-57-0
  • Molecular Weight : 159.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 0.8842/20
Country of Origin USA
Boiling point 131
Product Link http://www.matrixscientific.com/007558.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/007558.html
Availability details for
Quantity
  • Availability
  • Add to Cart