MAT047025783 Matrix Scientific

4-(2-Diethylaminoethyl)pyridine

  • MDL number MFCD00023637
  • Cas Number : 67580-61-4
  • Molecular Weight : 178.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 80-81�5 Torr
Product Link http://www.matrixscientific.com/007562.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007562.html
Availability details for
Quantity
  • Availability
  • Add to Cart