MAT047025784 Matrix Scientific

5-(3-Fluorophenyl)isoxazole

  • MDL number MFCD02183543
  • Cas Number : 387358-55-6
  • Molecular Weight : 163.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007026.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007026.html
Availability details for
Quantity
  • Availability
  • Add to Cart