MAT047025797 Matrix Scientific

6-(Dimethylamino)hexylamine

  • MDL number MFCD00047980
  • Cas Number : 1938-58-5
  • Molecular Weight : 144.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 0.831/25
Storage temp. Ambient
Country of Origin USA
Boiling point 67-68�Torr
Product Link http://www.matrixscientific.com/007575.html

Safety Information

hcodes Corrosive/Refrigerate
MSDS http://www.matrixscientific.com/007575.html
Availability details for
Quantity
  • Availability
  • Add to Cart