MAT047025827 Matrix Scientific

7-Hydroxyquinoline

  • MDL number MFCD00016730
  • Cas Number : 580-20-1
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/007603.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007603.html
Availability details for
Quantity
  • Availability
  • Add to Cart