MAT047025833 Matrix Scientific

3-Amino-2-cyano-5-(4-fluorophenyl)-thiophene

  • MDL number MFCD00210132
  • Cas Number : 210356-63-1
  • Molecular Weight : 218.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199-200
Country of Origin USA
Product Link http://www.matrixscientific.com/001662.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001662.html
Availability details for
Quantity
  • Availability
  • Add to Cart