MAT047025845 Matrix Scientific

5-(3-Nitrophenyl)isoxazole

  • MDL number MFCD02183546
  • Cas Number : 126633-02-1
  • Molecular Weight : 190.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007032.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007032.html
Availability details for
Quantity
  • Availability
  • Add to Cart