MAT047025874 Matrix Scientific

1-Acetyl-7-amino-2,3-dihydro-(1H)-indole

  • MDL number MFCD00203537
  • Cas Number : 51501-31-6
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159-160
Country of Origin USA
Product Link http://www.matrixscientific.com/006989.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006989.html
Availability details for
Quantity
  • Availability
  • Add to Cart