MAT047025877 Matrix Scientific

Ethyl 2,4-difluorobenzoate

  • MDL number MFCD00153149
  • Cas Number : 108928-00-3
  • Molecular Weight : 186.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 65-67�mm
Product Link http://www.matrixscientific.com/001667.html

Safety Information

hcodes Flammable
MSDS http://www.matrixscientific.com/001667.html
Availability details for
Quantity
  • Availability
  • Add to Cart