MAT047025889 Matrix Scientific

2,5-Bis(hexyl)-1,4-dibromobenzene

  • MDL number MFCD01318993
  • Cas Number : 117635-21-9
  • Molecular Weight : 404.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Fridge
Country of Origin USA
Product Link http://www.matrixscientific.com/007670.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/007670.html
Availability details for
Quantity
  • Availability
  • Add to Cart