MAT047025897 Matrix Scientific

4-(tert-Butoxymethyl)phenylboronic acid

  • MDL number MFCD03701685
  • Cas Number : 1024017-53-5
  • Molecular Weight : 208.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007678.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/007678.html
Availability details for
Quantity
  • Availability
  • Add to Cart