MAT047025899 Matrix Scientific

Ethyl 1,4-dihydro-8-fluoro-4-oxoquinoline-3-carboxylate

  • MDL number MFCD00052242
  • Cas Number : 71083-06-2
  • Molecular Weight : 235.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178
Country of Origin USA
Product Link http://www.matrixscientific.com/001669.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001669.html
Availability details for
Quantity
  • Availability
  • Add to Cart