MAT047025908 Matrix Scientific

5-(4-Nitrophenyl)isoxazole

  • MDL number MFCD02183547
  • Cas Number : 3383-42-4
  • Molecular Weight : 190.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176-178
Country of Origin USA
Product Link http://www.matrixscientific.com/007033.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007033.html
Availability details for
Quantity
  • Availability
  • Add to Cart