MAT047025910 Matrix Scientific

Ethyl 1,3-dimethyl-4-(4-fluorophenoxy)-1H-pyrazolo[3,4-b]pyridine-5-carboxylate

  • MDL number MFCD00067965
  • Cas Number : 174842-35-4
  • Molecular Weight : 329.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-111.5
Country of Origin USA
Product Link http://www.matrixscientific.com/001670.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001670.html
Availability details for
Quantity
  • Availability
  • Add to Cart