MAT047025914 Matrix Scientific

4-Nitrophenylboronic acid neopentylglycol ester

  • MDL number MFCD03701688
  • Cas Number : 502622-85-7
  • Molecular Weight : 235.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin USA
Product Link http://www.matrixscientific.com/007702.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/007702.html
Availability details for
Quantity
  • Availability
  • Add to Cart