MAT047025919 Matrix Scientific

4-(3-Pyridinyl)benzoic acid

  • MDL number MFCD03426513
  • Cas Number : 4385-75-5
  • Molecular Weight : 199.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/007710.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/007710.html
Availability details for
Quantity
  • Availability
  • Add to Cart