MAT047025933 Matrix Scientific

4-Hydroxy-1-methyl-2-oxo-1,2-dihydroquinoline-3-carboxylic acid (2-aminopheyl)amide

  • MDL number MFCD00629253
  • Cas Number : 151449-78-4
  • Molecular Weight : 309.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230-232
Country of Origin USA
Product Link http://www.matrixscientific.com/007730.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007730.html
Availability details for
Quantity
  • Availability
  • Add to Cart