MAT047025943 Matrix Scientific

4-Amino-3-ethylbenzonitrile

  • MDL number MFCD00041431
  • Cas Number : 170230-87-2
  • Molecular Weight : 146.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58-61
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/001673.html

Safety Information

hcodes Irritant/Irritant-Harmful
MSDS http://www.matrixscientific.com/001673.html
Availability details for
Quantity
  • Availability
  • Add to Cart